missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(chloromethyl) Sulfide 97.0+%, TCI America™

$122.44 - $1177.10

Chemical Identifiers

CAS 3592-44-7
Molecular Formula C2H4Cl2S
Molecular Weight (g/mol) 131.01
MDL Number MFCD00059474
InChI Key KRIFIIWBVJKVST-UHFFFAOYSA-N
Synonym Chloromethyl Sulfide, Dichlorodimethyl Sulfide, Di(chloromethyl) Sulfide
PubChem CID 565560
IUPAC Name chloro[(chloromethyl)sulfanyl]methane
SMILES ClCSCCl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B08431G
View Documents
TCI America
B08431G
1 g
Each for $122.44
 
B08435G
View Documents
TCI America
B08435G
5 g
Each for $406.86
 
B084325G
View Documents
TCI America
B084325G
25 g
Each for $1,177.10
 
Description

Description

Specifications

Chemical Identifiers

3592-44-7
131.01
KRIFIIWBVJKVST-UHFFFAOYSA-N
565560
ClCSCCl
C2H4Cl2S
MFCD00059474
Chloromethyl Sulfide, Dichlorodimethyl Sulfide, Di(chloromethyl) Sulfide
chloro[(chloromethyl)sulfanyl]methane

Specifications

3592-44-7
156°C
MFCD00059474
1992
KRIFIIWBVJKVST-UHFFFAOYSA-N
chloro[(chloromethyl)sulfanyl]methane
565560
≥97.0% (GC)
Bis(chloromethyl) Sulfide
Colorless
C2H4Cl2S
1 g
Chloromethyl Sulfide, Dichlorodimethyl Sulfide, Di(chloromethyl) Sulfide
ClCSCCl
131.01
131.01
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'