Learn More
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0), Pd 16.7%, Thermo Scientific Chemicals
$269.60 - $854.11
Chemical Identifiers
CAS | 1233717-68-4 |
---|---|
Molecular Formula | C32H56N2P2Pd |
Molecular Weight (g/mol) | 637.182 |
MDL Number | MFCD15071402 |
InChI Key | SSPOQURGNAWORH-UHFFFAOYSA-N |
Synonym | bis di-tert-butyl 4-dimethylaminophenyl phosphine palladium 0, bis 4-n,n-dimethylamino phenyl di-t-butylphosphino palladium 0, bis 4-di-tert-butylphosphanyl-n,n-dimethylaniline palladium |
PubChem CID | 46900632 |
IUPAC Name | 4-ditert-butylphosphanyl-N,N-dimethylaniline;palladium |
SMILES | CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.[Pd] |
Description
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0) is used in cross-coupling including sp2-sp3 coupling, especially Suzuki, Negishi-type, Heck, Sonogashira, and Buchwald-Hartwig coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1233717-68-4 | |
637.182 | |
SSPOQURGNAWORH-UHFFFAOYSA-N | |
46900632 | |
CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.[Pd] |
C32H56N2P2Pd | |
MFCD15071402 | |
bis di-tert-butyl 4-dimethylaminophenyl phosphine palladium 0, bis 4-n,n-dimethylamino phenyl di-t-butylphosphino palladium 0, bis 4-di-tert-butylphosphanyl-n,n-dimethylaniline palladium | |
4-ditert-butylphosphanyl-N,N-dimethylaniline;palladium |
Specifications
1233717-68-4 | |
C32H56N2P2Pd | |
250 mg | |
bis di-tert-butyl 4-dimethylaminophenyl phosphine palladium 0, bis 4-n,n-dimethylamino phenyl di-t-butylphosphino palladium 0, bis 4-di-tert-butylphosphanyl-n,n-dimethylaniline palladium | |
SSPOQURGNAWORH-UHFFFAOYSA-N | |
4-ditert-butylphosphanyl-N,N-dimethylaniline;palladium | |
46900632 | |
Pd 16.7% | |
Bis[di-tert-butyl (4-dimethylaminophenyl)phosphine]palladium (0) |
White | |
MFCD15071402 | |
Air Sensitive | |
Insoluble in water. | |
CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.[Pd] | |
637.182 | |
637.18 | |
Crystalline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only