Learn More
Bis(diisopropylamino)chlorophosphine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 321560050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bis(diisopropylamino)chlorophosphine | |
100.0°C to 110.0°C | |
C12H28ClN2P | |
MFCD00061482 | |
bis diisopropylamino chlorophosphine, phosphorodiamidous chloride, tetrakis 1-methylethyl, chloro diisopropylamino phosphanyl diisopropylamine, pubchem6478, acmc-1av77, bisdiisopropylaminochlorophosphine, bis diisopropyamino chlorophosphine, bis di-isopropylamino chlorophosphine, bis n,n-diisopropylamino chlorophosphine, chlorobis n,n-diisopropylamino phosphine | |
FEHUTHGOLLQBNW-UHFFFAOYSA-N | |
N-[chloro-[di(propan-2-yl)amino]phosphanyl]-N-propan-2-ylpropan-2-amine | |
2733425 |
56183-63-2 | |
Glass bottle | |
[((CH3)2CH)2N]2PCl | |
5 g | |
Solubility in water: reacts | |
CC(C)N(C(C)C)P(Cl)N(C(C)C)C(C)C | |
266.79 | |
266.8 |
Chemical Identifiers
56183-63-2 | |
266.79 | |
FEHUTHGOLLQBNW-UHFFFAOYSA-N | |
2733425 | |
CC(C)N(C(C)C)P(Cl)N(C(C)C)C(C)C |
C12H28ClN2P | |
MFCD00061482 | |
bis diisopropylamino chlorophosphine, phosphorodiamidous chloride, tetrakis 1-methylethyl, chloro diisopropylamino phosphanyl diisopropylamine, pubchem6478, acmc-1av77, bisdiisopropylaminochlorophosphine, bis diisopropyamino chlorophosphine, bis di-isopropylamino chlorophosphine, bis n,n-diisopropylamino chlorophosphine, chlorobis n,n-diisopropylamino phosphine | |
N-[chloro-[di(propan-2-yl)amino]phosphanyl]-N-propan-2-ylpropan-2-amine |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
RUO – Research Use Only