missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(pinacolato)diboron, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 330570250

 View more versions of this product

Catalog No. AC330570250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bis(pinacolato)diboron
135.0°C to 140.0°C
Authentic
Glass bottle
MFCD00799570
15,1301
IPWKHHSGDUIRAH-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
2733548
98%
73183-34-3
White
98%
C12H24B2O4
25 g
bis pinacolato diboron, bis pinacolato diborane, 4,4,4',4',5,5,5',5'-octamethyl-2,2'-bi 1,3,2-dioxaborolane, pinacol diborane, diboron pinacol ester, b2pin2, 4,4,4',4',5,5,5',5'-octamethyl-2,2'-bi-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3,2-dioxaborolane, bispinacolate diboron, unii-i906w26p4u
CC1(C)OB(OC1(C)C)B1OC(C)(C)C(C)(C)O1
253.94
253.9
Powder or Platelets

Chemical Identifiers

73183-34-3
253.94
IPWKHHSGDUIRAH-UHFFFAOYSA-N
2733548
CC1(C)OB(OC1(C)C)B1OC(C)(C)C(C)(C)O1
C12H24B2O4
MFCD00799570
bis pinacolato diboron, bis pinacolato diborane, 4,4,4',4',5,5,5',5'-octamethyl-2,2'-bi 1,3,2-dioxaborolane, pinacol diborane, diboron pinacol ester, b2pin2, 4,4,4',4',5,5,5',5'-octamethyl-2,2'-bi-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3,2-dioxaborolane, bispinacolate diboron, unii-i906w26p4u
4,4,5,5-tetramethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only