missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(tri-tert-butylphosphine)palladium(0), Thermo Scientific Chemicals

Coupling reactions, Heck couplings

$225.79 - $764.32

Chemical Identifiers

CAS 53199-31-8
Molecular Formula C24H54P2Pd
Molecular Weight (g/mol) 511.06
MDL Number MFCD03094580
InChI Key MXQOYLRVSVOCQT-UHFFFAOYSA-N
Synonym bis tri-tert-butylphosphine palladium 0, bis tri-t-butylphosphine palladium 0, bis tri-tert-butylphosphine palladium, bis tri-tert-butylphosphane palladium, palladium; tritert-butylphosphane, palladium, bis tris 1,1-dimethylethyl phosphine, pd p tbu 3 2, pd t-bu3p 2, bis tri-t-butylphosphine palladium, di tri-tert-butylphosphine palladium 0
PubChem CID 2734558
IUPAC Name palladium;tritert-butylphosphane
SMILES [Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4484501
View Documents
Thermo Scientific Chemicals
04484501
0.25 g
Each for $225.79
 
AA4484503
View Documents
Thermo Scientific Chemicals
04484503
1 g
Each for $764.32
 
Description

Description

Bis(tri-tert-butylphosphine)palladium(0) is used in Coupling reactions, Heck couplings. It acts as a palladium catalyst for reductive carbonylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under inert atmosphere
Specifications

Chemical Identifiers

53199-31-8
511.06
MXQOYLRVSVOCQT-UHFFFAOYSA-N
2734558
[Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C
C24H54P2Pd
MFCD03094580
bis tri-tert-butylphosphine palladium 0, bis tri-t-butylphosphine palladium 0, bis tri-tert-butylphosphine palladium, bis tri-tert-butylphosphane palladium, palladium; tritert-butylphosphane, palladium, bis tris 1,1-dimethylethyl phosphine, pd p tbu 3 2, pd t-bu3p 2, bis tri-t-butylphosphine palladium, di tri-tert-butylphosphine palladium 0
palladium;tritert-butylphosphane

Specifications

53199-31-8
White
MFCD03094580
Air and light sensitive
Soluble in toluene benzene,slight soluble in chloroform and acetone. Decomposes in water.
[Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C
511.06
511.06
Powder
258°C to 272°C
C24H54P2Pd
0.25 g
bis tri-tert-butylphosphine palladium 0, bis tri-t-butylphosphine palladium 0, bis tri-tert-butylphosphine palladium, bis tri-tert-butylphosphane palladium, palladium; tritert-butylphosphane, palladium, bis tris 1,1-dimethylethyl phosphine, pd p tbu 3 2, pd t-bu3p 2, bis tri-t-butylphosphine palladium, di tri-tert-butylphosphine palladium 0
MXQOYLRVSVOCQT-UHFFFAOYSA-N
palladium;tritert-butylphosphane
2734558
Pd 20.9%
Bis (tri-tert-butylphosphine)palladium (0)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 :

  • Packaged under inert atmosphere

Recommended Storage : Store at -20°C; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only