missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(trimethylsilyl) Sulfide 97.0+%, TCI America™
$283.33 - $1017.78
Chemical Identifiers
| CAS | 3385-94-2 |
|---|---|
| Molecular Formula | C6H18SSi2 |
| Molecular Weight (g/mol) | 178.44 |
| MDL Number | MFCD00014851 |
| InChI Key | RLECCBFNWDXKPK-UHFFFAOYSA-N |
| Synonym | bis trimethylsilyl sulfide, hexamethyldisilthiane, hexamethyldisilathiane, disilathiane, hexamethyl, 1,1,1,3,3,3-hexamethyldisilathiane, disilthiane, hexamethyl, disilathiane, 1,1,1,3,3,3-hexamethyl, thiobis trimethylsilane, acmc-1ckj8 |
| PubChem CID | 76920 |
| IUPAC Name | trimethyl(trimethylsilylsulfanyl)silane |
| SMILES | C[Si](C)(C)S[Si](C)(C)C |
Chemical Identifiers
| 3385-94-2 | |
| 178.44 | |
| RLECCBFNWDXKPK-UHFFFAOYSA-N | |
| 76920 | |
| C[Si](C)(C)S[Si](C)(C)C |
| C6H18SSi2 | |
| MFCD00014851 | |
| bis trimethylsilyl sulfide, hexamethyldisilthiane, hexamethyldisilathiane, disilathiane, hexamethyl, 1,1,1,3,3,3-hexamethyldisilathiane, disilthiane, hexamethyl, disilathiane, 1,1,1,3,3,3-hexamethyl, thiobis trimethylsilane, acmc-1ckj8 | |
| trimethyl(trimethylsilylsulfanyl)silane |
Specifications
| 3385-94-2 | |
| 163°C | |
| MFCD00014851 | |
| 1993 | |
| RLECCBFNWDXKPK-UHFFFAOYSA-N | |
| trimethyl(trimethylsilylsulfanyl)silane | |
| 76920 | |
| ≥97.0% (GC) | |
| Bis(trimethylsilyl) Sulfide |
| Yellow | |
| C6H18SSi2 | |
| 5 g | |
| bis trimethylsilyl sulfide, hexamethyldisilthiane, hexamethyldisilathiane, disilathiane, hexamethyl, 1,1,1,3,3,3-hexamethyldisilathiane, disilthiane, hexamethyl, disilathiane, 1,1,1,3,3,3-hexamethyl, thiobis trimethylsilane, acmc-1ckj8 | |
| C[Si](C)(C)S[Si](C)(C)C | |
| 178.44 | |
| 178.44 | |
| Liquid |
Safety and Handling
TSCA : Yes