missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(trimethylsilyl)trifluoroacetamide, BSTFA, EMD-Millipore
Supplier: Technidata 1102550005
Specifications
Bis(trimethylsilyl)trifluoroacetamide | |
Glass | |
C8H18F3NOSi2 | |
XCOBLONWWXQEBS-GHXNOFRVSA-N | |
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate | |
9601896 | |
Density: 0.97g/cm3 (20°C) |
25561-30-2 | |
5 mL | |
2,2,2-Trifluoro-N,O-bis(trimethylsilyl)acetamide | |
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C | |
257.403 | |
257.4 |
Chemical Identifiers
25561-30-2 | |
257.403 | |
2,2,2-Trifluoro-N,O-bis(trimethylsilyl)acetamide | |
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate |
C8H18F3NOSi2 | |
XCOBLONWWXQEBS-GHXNOFRVSA-N | |
9601896 | |
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C |
Safety and Handling
Recommended Storage : Store at 15° to 25°C