missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(trimethylsilyl)trifluoroacetamide, BSTFA, EMD-Millipore

Supplier:  Technidata 1102550025

 View more versions of this product

Catalog No. M1102550025


Explore available promotions

Description

Description

Specifications

Specifications

Bis(trimethylsilyl)trifluoroacetamide
Glass
C8H18F3NOSi2
XCOBLONWWXQEBS-GHXNOFRVSA-N
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
9601896
Density: 0.97g/cm3 (20°C)
25561-30-2
25 mL
2,2,2-Trifluoro-N,O-bis(trimethylsilyl)acetamide
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
257.403
257.4

Chemical Identifiers

25561-30-2
257.403
2,2,2-Trifluoro-N,O-bis(trimethylsilyl)acetamide
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
C8H18F3NOSi2
XCOBLONWWXQEBS-GHXNOFRVSA-N
9601896
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 15° to 25°C

SDS
Documents

Documents

Promotions

Promotions