missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis-Tris, ≥99%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219454691

 View more versions of this product

Catalog No. ICN19454691


Explore available promotions

Description

Description

Specifications

Specifications

Bis-Tris
6976-37-0
Crystalline Powder
C8H19NO5
2,2-bis(Hydroxymethyl)-2,2',2''-nitrilotriethanol, 2, 2-bis(Hydroxymethyl)-2, 2'
C(CO)N(CCO)C(CO)(CO)CO
209.242
CHEBI:41250
≥99%
Cell culture reagent
White
1 kg
MFCD00002853
OWMVSZAMULFTJU-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
81462
209.2

Chemical Identifiers

6976-37-0
209.242
OWMVSZAMULFTJU-UHFFFAOYSA-N
81462
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
C8H19NO5
MFCD00002853
2,2-bis(Hydroxymethyl)-2,2',2''-nitrilotriethanol, 2, 2-bis(Hydroxymethyl)-2, 2'
CHEBI:41250
C(CO)N(CCO)C(CO)(CO)CO
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature (15-30°C).

SDS
Documents

Documents

Promotions

Promotions