Learn More
Bismuth 2-ethylhexanoate, Thermo Scientific Chemicals
$55.37 - $489.19
Chemical Identifiers
CAS | 67874-71-9 |
---|---|
Molecular Formula | C24H48BiO6 |
Molecular Weight (g/mol) | 641.622 |
MDL Number | MFCD00043803 |
InChI Key | MWXWNYHYPMRCHN-UHFFFAOYSA-N |
Synonym | 2-Ethylhexanoic acid bismuth salt |
PubChem CID | 131857451 |
IUPAC Name | bismuth;2-ethylhexanoic acid |
SMILES | CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA3671006
|
Thermo Scientific Chemicals
03671006 |
5 g |
Each for $55.37
|
|
|||||
AA3671014
|
Thermo Scientific Chemicals
03671014 |
25 g |
Each for $167.27
|
|
|||||
AA3671022
|
Thermo Scientific Chemicals
03671022 |
100 g |
Each for $489.19
|
|
|||||
Description
Bismuth 2-ethylhexanoate is used as an industrial lubricant and dessicant. It is also used for synthesizing of urethane foam, urethane coating and other urethane products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67874-71-9 | |
641.622 | |
MWXWNYHYPMRCHN-UHFFFAOYSA-N | |
131857451 | |
CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi] |
C24H48BiO6 | |
MFCD00043803 | |
2-Ethylhexanoic acid bismuth salt | |
bismuth;2-ethylhexanoic acid |
Specifications
67874-71-9 | |
MFCD00043803 | |
2-Ethylhexanoic acid bismuth salt | |
MWXWNYHYPMRCHN-UHFFFAOYSA-N | |
bismuth;2-ethylhexanoic acid | |
131857451 | |
Viscous Liquid |
C24H48BiO6 | |
5 g | |
Not miscible with water. | |
CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi] | |
641.622 | |
638.61 | |
Bismuth 2-ethylhexanoate |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H361d
EINECSNumber : 267-499-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only