Learn More
Bismuth hydroxide, tech., Thermo Scientific Chemicals
Hydrolysis of ribonucleic acid, absorbent
$204.53 - $677.51
Chemical Identifiers
CAS | 10361-43-0 |
---|---|
Molecular Formula | BiH3O3 |
Molecular Weight (g/mol) | 260.00 |
MDL Number | MFCD00046188 |
InChI Key | TZSXPYWRDWEXHG-UHFFFAOYSA-K |
IUPAC Name | bismuth(3+) trihydroxide |
SMILES | [OH-].[OH-].[OH-].[Bi+3] |
Description
It is used for the hydrolysis of ribonucleic acid and as absorbent for rutin and quercetin. It is a component used in milk of bismuth is used in gastrointestinal disorders as a protective agent. It is used for the removal of nitrate and other anions from water by yellow bismuth hydroxide. A bismuth oxide nanoplate is useful for carbon dioxide gas sensor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10361-43-0 | |
260.00 | |
TZSXPYWRDWEXHG-UHFFFAOYSA-K | |
[OH-].[OH-].[OH-].[Bi+3] |
BiH3O3 | |
MFCD00046188 | |
bismuth(3+) trihydroxide |
Specifications
10361-43-0 | |
250 g | |
MFCD00046188 | |
TZSXPYWRDWEXHG-UHFFFAOYSA-K | |
bismuth(3+) trihydroxide | |
260 | |
Odorless |
Amorphous Powder | |
BiH3O3 | |
Insoluble in water; Soluble in acids. | |
[OH-].[OH-].[OH-].[Bi+3] | |
260.00 | |
Technical | |
Bismuth hydroxide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only