missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth(III) bromide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 447770250

 View more versions of this product

Catalog No. AC447770250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

453°C
218°C
25 g
BiBr3
15,1262
Solubility in water: reacts. Other solubilities: practically insoluble in alcohol,soluble in solutions of ki,kbr,kcl,dil hcl
Br[Bi](Br)Br
448.69
448.69
Glass bottle
Bismuth(III) bromide
7787-58-8
96 to 103% (Bi)
BiBr3
bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo
TXKAQZRUJUNDHI-UHFFFAOYSA-K
tribromobismuthane
82232
97%

Chemical Identifiers

7787-58-8
448.69
bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo
tribromobismuthane
BiBr3
TXKAQZRUJUNDHI-UHFFFAOYSA-K
82232
Br[Bi](Br)Br
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 232-121-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only