missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth(III) iodide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01065609

 View more versions of this product

Catalog No. AA1065609


Explore available promotions

Description

Description

Polycrystalline films of bismuth triiodide are used as beta direct and digital detectors for medical imaging applications. Bismuth(III) iodide is used as an ice nucleant. It is also used in electron microscopy to reveal synapses. Addition of bismuth triiodide has been shown to improve the glass forming ability and thermal stability of Ge-Te-BiI(3) chalcogenide glasses. Solution-processable BiI3 nanosheets have been shown to exhibit the potential to be used as HTLs (Hole Transport Layer) in conventional OPV (organic photovoltaic) devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

∼500°C
408°C
Gray to Black
10 g
BiI3
UN3260
bismuth iodide, bismuth iii iodide, bismuth triiodide, bismuthine, triiodo, triiodobismuthine, triiodobismuth, bismuth iodide,, ksc179g5d
KOECRLKKXSXCPB-UHFFFAOYSA-K
bismuth(3+) triiodide
111042
99.999%
Acidic
5.778 g/mL
Bismuth(III) iodide
7787-64-6
Lump/Powder
(metals basis)
MFCD00010894
14,1269
Insoluble in water. Highly soluble in ethanol,benzene,toluene and xylene,hydrochloric acid and in hydriodic acid.
[I-].[I-].[I-].[Bi+3]
589.69
589.69
Puratronic™
Hygroscopic; Light sensitive

Chemical Identifiers

7787-64-6
589.69
KOECRLKKXSXCPB-UHFFFAOYSA-K
111042
BiI3
MFCD00010894
bismuth iodide, bismuth iii iodide, bismuth triiodide, bismuthine, triiodo, triiodobismuthine, triiodobismuth, bismuth iodide,, ksc179g5d
[I-].[I-].[I-].[Bi+3]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 232-127-4

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only