Learn More
Bismuth(III) isopropoxide, Thermo Scientific Chemicals
$911.11 - $911.11
Chemical Identifiers
CAS | 15049-67-9 |
---|---|
Molecular Formula | C9H21BiO3 |
Molecular Weight (g/mol) | 386.244 |
MDL Number | MFCD00799065 |
InChI Key | KNPRLIQQQKEOJN-UHFFFAOYSA-N |
Synonym | bismuth iii isopropoxide, triisopropoxybismuthine, acmc-20all6, bismuth 3+ tripropan-2-olate, bismuth 3+ tris propan-2-olate |
PubChem CID | 22648700 |
IUPAC Name | bismuth;propan-2-olate |
SMILES | CC(C)[O-].CC(C)[O-].CC(C)[O-].[Bi+3] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4393806
|
Thermo Scientific Chemicals
04393806 |
5 g |
Each for $911.11
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
15049-67-9 | |
386.244 | |
KNPRLIQQQKEOJN-UHFFFAOYSA-N | |
22648700 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Bi+3] |
C9H21BiO3 | |
MFCD00799065 | |
bismuth iii isopropoxide, triisopropoxybismuthine, acmc-20all6, bismuth 3+ tripropan-2-olate, bismuth 3+ tris propan-2-olate | |
bismuth;propan-2-olate |
Specifications
15049-67-9 | |
C9H21BiO3 | |
MFCD00799065 | |
UN3181 | |
bismuth iii isopropoxide, triisopropoxybismuthine, acmc-20all6, bismuth 3+ tripropan-2-olate, bismuth 3+ tris propan-2-olate | |
KNPRLIQQQKEOJN-UHFFFAOYSA-N | |
bismuth;propan-2-olate | |
22648700 | |
Powder |
Gray | |
Bi[OCH(CH3)2]3 | |
5 g | |
Moisture sensitive | |
Reacts with water. | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Bi+3] | |
386.244 | |
386.24 | |
Bismuth (III) isopropoxide |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA : No
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only