missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth(III) nitrate pentahydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1174822

 View more versions of this product

Catalog No. AAA1174822


Explore available promotions

Description

Description

Bismuth(III) nitrate pentahydrate is an oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines. It also serves as a reagent for selective oxidation of sufides to sulfoxides. Further, it is involved in the preparation of Dragendorff reagent and used as a TLC stain. It is also used to prepare other bismuth compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bismuth(III) nitrate pentahydrate is an oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines. It also serves as a reagent for selective oxidation of sufides to sulfoxides. Further, it is involved in the preparation of Dragendorff reagent and used as a TLC stain. It is also used to prepare other bismuth compounds.

Solubility
Soluble in dilute nitric acid solution, dilute acetic acid, glycerol and acetone. Insoluble in alcohol, ethyl acetateSoluble in acetone, acetic acid and glycerol. Insoluble in ethanol and ethyl acetate.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong acids, strong reducing agents, strong oxidizing agents, powdered metals and organic materials.
Specifications

Specifications

Bismuth (III) nitrate pentahydrate
10035-06-0
BiH10N3O14
UN1477
bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate
FBXVOTBTGXARNA-UHFFFAOYSA-N
bismuth;trinitrate;pentahydrate
160911
98%
Hygroscopic
30°C (decomposition)
100 g
MFCD00149157
14,1271
Soluble in dilute nitric acid solution,dilute acetic acid,glycerol and acetone. Insoluble in alcohol,ethyl acetateSoluble in acetone,acetic acid and glycerol. Insoluble in ethanol and ethyl acetate.
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
485.07
485.07 (394.99 Anhydrous)
Odorless
2.83

Chemical Identifiers

10035-06-0
485.07
FBXVOTBTGXARNA-UHFFFAOYSA-N
160911
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
BiH10N3O14
MFCD00149157
bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate
bismuth;trinitrate;pentahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P220-P221-P261-P262-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335-H373

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

EINECSNumber : 233-791-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only