missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth(III) sulfate, 99%, Thermo Scientific Chemicals

$65.21 - $211.84

Chemical Identifiers

CAS 7787-68-0
Molecular Formula Bi2O12S3
Molecular Weight (g/mol) 706.13
MDL Number MFCD00036140
InChI Key BEQZMQXCOWIHRY-UHFFFAOYSA-H
Synonym bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4
PubChem CID 198144
IUPAC Name dibismuth;trisulfate
SMILES [Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2200714
View Documents
Thermo Scientific Chemicals
B2200714
25 g
Each for $65.21
 
AAB2200722
View Documents
Thermo Scientific Chemicals
B2200722
100 g
Each for $211.84
 
Description

Description

The esterification of thujopsene with bismuth(III) sulfate in carboxylic acids yeilds esters in 30%. Bismuth(III) sulfateis used as analytics reagents and medicine, it is also used in pharmaceutical industry. It is used to determine other metal sulphate salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The esterification of thujopsene with bismuth(III) sulfate in carboxylic acids yeilds esters in 30%. Bismuth(III) sulfateis used as analytics reagents and medicine, it is also used in pharmaceutical industry. It is used to determine other metal sulphate salts.

Solubility
Soluble in acid, insoluble in water and ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

7787-68-0
706.13
BEQZMQXCOWIHRY-UHFFFAOYSA-H
198144
[Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Bi2O12S3
MFCD00036140
bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4
dibismuth;trisulfate

Specifications

405°C (decomposition)
25 g
Bi2(SO4)3
14,1286
Soluble in acid,insoluble in water and ethanol.
[Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
706.13
706.13
5.08
7787-68-0
Bi2O12S3
MFCD00036140
bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4
BEQZMQXCOWIHRY-UHFFFAOYSA-H
dibismuth;trisulfate
198144
99%
Bismuth(III) sulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 232-129-5

RTECSNumber : WS6600000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only