missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bismuth Nitrate, Pentahydrate, Crystal, ACS, 98%, Spectrum™ Chemical
$296.44 - $13192.32
Chemical Identifiers
CAS | 10035-06-0 |
---|---|
Molecular Formula | BiH10N3O14 |
Molecular Weight (g/mol) | 485.07 |
InChI Key | FBXVOTBTGXARNA-UHFFFAOYSA-N |
IUPAC Name | bismuth(3+) pentahydrate trinitrate |
SMILES | O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
18609100
|
Spectrum Chemical Mfg Cor
B111525GM |
25 g | Amber Glass Bottle |
Each for $296.44
|
|
||||
18609101
|
Spectrum Chemical Mfg Cor
B1115125GM |
125 g | Amber Glass Bottle |
Each for $562.21
|
|
||||
18609102
|
Spectrum Chemical Mfg Cor
B1115500GM |
500 g | Amber Glass Bottle |
Each for $1,247.74
|
|
||||
18609103
|
Spectrum Chemical Mfg Cor
B111525KG |
2.5 kg | Amber Glass Bottle |
Each for $3,898.18
|
|
||||
18609104
|
Spectrum Chemical Mfg Cor
B111512KG |
12 kg | Poly Pail |
Each for $6,873.53
|
|
||||
18609105
|
Spectrum Chemical Mfg Cor
B111550KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
10035-06-0 | |
485.07 | |
bismuth(3+) pentahydrate trinitrate |
BiH10N3O14 | |
FBXVOTBTGXARNA-UHFFFAOYSA-N | |
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
10035-06-0 | |
25 g | |
FBXVOTBTGXARNA-UHFFFAOYSA-N | |
bismuth(3+) pentahydrate trinitrate | |
98% | |
Amber Glass Bottle |
1 | |
BiH10N3O14 | |
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
485.07 | |
ACS |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.