missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bismuth Subcarbonate, Purified, Spectrum™ Chemical
$350.59 - $30272.59
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18609106
|
Spectrum Chemical Mfg Cor
BI140125GM |
125 g | Amber Glass Bottle |
Each for $350.59
|
|
||||
18609107
|
Spectrum Chemical Mfg Cor
BI140500GM |
500 g | Amber Glass Bottle |
Each for $917.29
|
|
||||
18609108
|
Spectrum Chemical Mfg Cor
BI14025KG |
2.5 kg | Amber Glass Bottle |
Each for $3,203.65
|
|
||||
18609109
|
Spectrum Chemical Mfg Cor
BI14012KG |
12 kg | Poly Pail |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18609110
|
Spectrum Chemical Mfg Cor
BI14045KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
5892-10-4 |
Specifications
5892-10-4 | |
Amber Glass Bottle | |
Purified |
1 | |
125 g |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.