missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bisphenol A diglycidyl ether resin, Thermo Scientific Chemicals

$55.88 - $374.37

Chemical Identifiers

CAS 1675-54-3
Molecular Formula C21H24O4
Molecular Weight (g/mol) 340.419
MDL Number MFCD00080480
InChI Key LCFVJGUPQDGYKZ-UHFFFAOYSA-N
Synonym bisphenol a diglycidyl ether, 2,2-bis 4-glycidyloxyphenyl propane, epoxide a, epophen el 5, badge, dian diglycidyl ether, diglycidyl bisphenol a, epi-rez 510, bpdge, dgeba
PubChem CID 2286
ChEBI CHEBI:34578
IUPAC Name 2-[[4-[2-[4-(oxiran-2-ylmethoxy)phenyl]propan-2-yl]phenoxy]methyl]oxirane
SMILES CC(C)(C1=CC=C(C=C1)OCC2CO2)C3=CC=C(C=C3)OCC4CO4
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1481714
View Documents
Thermo Scientific Chemicals
L1481714
25 g
Each for $55.88
 
AAL1481722
View Documents
Thermo Scientific Chemicals
L1481722
100 g
Each for $122.18
 
AAL1481736
View Documents
Thermo Scientific Chemicals
L1481736
500 g
Each for $374.37
 
Description

Description

Bisphenol A diglycidyl ether is used to inhibit the peroxisome proliferator-activated receptor (PPARγ) and adipocyte differentiation and it induce apoptosis in tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bisphenol A diglycidyl ether is used to inhibit the peroxisome proliferator-activated receptor (PPARγ) and adipocyte differentiation and it induce apoptosis in tumor cells.

Solubility
Soluble in 100% ethanol, dimethyl sulfoxide (100 mM), dimethyl formamide, chloroform, methanol, and ethanol (50 mM). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

1675-54-3
340.419
LCFVJGUPQDGYKZ-UHFFFAOYSA-N
2286
2-[[4-[2-[4-(oxiran-2-ylmethoxy)phenyl]propan-2-yl]phenoxy]methyl]oxirane
C21H24O4
MFCD00080480
bisphenol a diglycidyl ether, 2,2-bis 4-glycidyloxyphenyl propane, epoxide a, epophen el 5, badge, dian diglycidyl ether, diglycidyl bisphenol a, epi-rez 510, bpdge, dgeba
CHEBI:34578
CC(C)(C1=CC=C(C=C1)OCC2CO2)C3=CC=C(C=C3)OCC4CO4

Specifications

1675-54-3
C21H24O4
MFCD00080480
299026
Soluble in 100% ethanol,dimethyl sulfoxide (100 mM),dimethyl formamide,chloroform,methanol,and ethanol (50 mM). Insoluble in water.
CC(C)(C1=CC=C(C=C1)OCC2CO2)C3=CC=C(C=C3)OCC4CO4
340.419
CHEBI:34578
Bisphenol A diglycidyl ether
1.17
1.573
25 g
bisphenol a diglycidyl ether, 2,2-bis 4-glycidyloxyphenyl propane, epoxide a, epophen el 5, badge, dian diglycidyl ether, diglycidyl bisphenol a, epi-rez 510, bpdge, dgeba
LCFVJGUPQDGYKZ-UHFFFAOYSA-N
2-[[4-[2-[4-(oxiran-2-ylmethoxy)phenyl]propan-2-yl]phenoxy]methyl]oxirane
2286
340.42
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c

H315-H317-H319

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 216-823-5

RTECSNumber : TX3800000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only