missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Biuret, 97%, extra pure

$37.81 - $262.19

Chemical Identifiers

CAS 108-19-0
Molecular Formula C2H5N3O2
Molecular Weight (g/mol) 103.08
InChI Key OHJMTUPIZMNBFR-UHFFFAOYSA-N
Synonym biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
PubChem CID 7913
ChEBI CHEBI:18138
IUPAC Name carbamoylurea
SMILES C(=O)(N)NC(=O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC106520050
SDS View Documents
Thermo Scientific Chemicals
106520050
5 g Glass bottle
Each for $37.81
 
AC106520250
SDS View Documents
Thermo Scientific Chemicals
106520250
25 g Glass bottle
Each for $110.32
 
AC106521000
SDS View Documents
Thermo Scientific Chemicals
106521000
100 g Glass bottle
Each for $262.19
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-19-0
103.08
biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
CHEBI:18138
C(=O)(N)NC(=O)N
C2H5N3O2
OHJMTUPIZMNBFR-UHFFFAOYSA-N
7913
carbamoylurea

Specifications

108-19-0
White
Authentic
Glass bottle
H2NCONHCONH2
03,7
biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
OHJMTUPIZMNBFR-UHFFFAOYSA-N
carbamoylurea
7913
103.08
Extra Pure
Biuret
187°C to 194°C
1% max.
96% min. (ex N)
C2H5N3O2
5 g
15,131
Solubility in water: 2g/100mL water (25°C). Other solubilities: freely soluble in alcohols,slightly soluble in diethyl ether
C(=O)(N)NC(=O)N
103.08
CHEBI:18138
97%
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-559-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only