Learn More
Thermo Scientific™ Black 96-Well Immuno Plates
Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with these black, 96-well immuno plates, available in a choice of three surface types.
$638.07 - $1609.63
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Detection Method | Fluorescence |
Array | 8 x 12 |
Color | Black |
Catalog Number | Mfr. No. | Surface Treatment | Volume (Metric) Working | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Surface Treatment | Volume (Metric) Working | Price | Quantity | ||||
12-566-24
|
Thermo Scientific™
437111 |
MaxiSorp | 350 μL |
Case of 80 Each for $1,536.28
|
|
||||
12-566-30
|
Thermo Scientific™
437112 |
PolySorp | 350 μL |
Case of 80 Each for $1,609.63
|
|
||||
14-245-197A
|
Thermo Scientific™
7605 |
Medium Binding | 280 μL |
Case of 50 Each for $638.07
|
|
||||
14-245-177
|
Thermo Scientific™
7805 |
High Binding | 280 μL |
Case of 50 Each for $778.36
|
|
||||
21-377-271
|
Thermo Scientific™
9502867 |
Universal Binding | 350 μL |
Case of 50 Each for $1,086.94
|
|
||||
Description
Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ Black 96-Well Immuno Plates. Featuring a choice of three surface types and optimal binding, these plates minimize background in fluorescence reading.
Features:
- Plate has SBS footprint — fits standard equipment
- Available with flat- or round-bottom wells
- Certified binding homogeneity/reproducibility
Choice of surface:
- Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) are for absorption of hydrophobic molecules.
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.
- Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules.
Compatible with:
Compatible with lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country
Specifications
Standard | |
Fluorescence | |
Black | |
Polystyrene | |
Round, Flat Bottom (F-well) | |
None |
Without Lid | |
8 x 12 | |
Microplate | |
96 | |
Fluorescence |