Fisherbrand™ Black Phenolic Work Surface
Phenolic Work Surface is a chemically resistant, one-inch thick black Phenolic top.
 View more versions of this product
Catalog No. 06000374
Explore available promotions
Description
Description
  • Front edge finished, all other edges are square
  • Curb must be ordered separately
Specifications
Specifications
2 Years
Work Surface
Work Surface
1 in.
48 in.
80 lb.
121.9 cm
None
Phenolic
None
30 in.
2.5 cm
Work Surface
37 kg
76.2 cm
Smooth, Non-glare Surface
Black