Fisherbrand™ Black Phenolic Work Surface
Phenolic Work Surface is a chemically resistant, one-inch thick black Phenolic top.
Description
- Front edge finished, all other edges are square
- Curb must be ordered separately
Specifications
2 Years | |
Work Surface | |
Work Surface | |
1 in. | |
72 in. | |
120 lb. | |
182.9 cm | |
None | |
Phenolic |
None | |
30 in. | |
2.5 cm | |
Work Surface | |
55 kg | |
76.2 cm | |
Smooth, Non-glare Surface | |
Black |