Black Stallion™ Aluminized Heat-Resistant Coats and Pants
Supplier:  Black Stallion™ 204AHSXLG
 View more versions of this product
Catalog No. 19-056851
Explore available promotions
Description
Description
  • Together, these aluminized garments protect trunk, arms and legs from radiant heat and molten splash
  • Coat, 30 in. long (76cm), features mandarin collar and snap-front closure
  • Also has inside breast pocket and set-in sleeves with open cuffs
  • Pants have reinforced waistband and flame-retardant hook-and-loop fly
  • 19 oz. aluinized carbon/Kevlar
Specifications
Specifications
42
Carbon Dupont™ Kevlar™ aramid fibers
Silver