Black Stallion™ Aluminized Heat-Resistant Coats and Pants
Supplier: Black Stallion™ 130AHSXXL
Description
- Together, these aluminized garments protect trunk, arms and legs from radiant heat and molten splash
- Coat, 30 in. long (76cm), features mandarin collar and snap-front closure
- Also has inside breast pocket and set-in sleeves with open cuffs
- Pants have reinforced waistband and flame-retardant hook-and-loop fly
- 19 oz. aluinized carbon/Kevlar
Specifications
52 | |
Carbon Dupont™ Kevlar™ aramid fibers | |
Silver |