FEATHER Blade for Replaceable Blade Scissors No.145
FEATHER Blades for Replaceable Blade Scissors No.145 come in three different blade types: sharp/sharp, sharp/blunt and blunt/blunt and can all be used with the same handle (sold separately). The replaceable blade design eliminates the cost and trouble of resharpening.
Supplier:  FEATHER 46BLBL
 View more versions of this product
Catalog No. 13-804-26
Explore available promotions
Description
Description
  • Made in Japan
  • Sharp/Sharp or Sharp/Blunt or Blunt/Blunt styles available
  • Stainless steel blades
Specifications
Specifications
Scissor Blades
2/Pk.
Replaceable Blade Scissors No. 145
48 mm
Stainless Steel