Mopec BladeFLASK Blade Remover, Qlicksmart
Qlicksmart™ Blade Removal System from Mopec is a medical safety device specially designed, engineered and manufactured to comply with recently revised OSHA Bloodborne Pathogens Standards.
Supplier:  Mopec AJ150
Catalog No. AJ150
Explore available promotions
Description
Description
  • Single-handed removal and immediate containment of contaminated scalpel blades which dramatically reduces the risk and cost of injury to the user
  • Built-in counter and automatic shut off when full
  • Holds 100 blades
  • Fits Blade #: 10, 10a, 11, 12, 15, 20, 21, 22, 23, 24
  • Fits Handle #: 3 (AJ140), 4 (AJ101), 4L (AJ102), 5, 7, 9
  • Easy Mounting with Optional AJ151 Mounting Bracket
  • OSHA Compliant
Specifications
Specifications
United States Version