Bullard™ Blower Assembly for (EVAHL) PAPR System
Designed to be used with the EVAHL Powered Air Purifying Respirator
Supplier:  Bullard™ EVAHL4
 View more versions of this product
Catalog No. 17-989-537
Explore available promotions
Description
Description
  • Choice of assembly configurations
Compliance
Meets NIOSH; ANSI™/ISA 2.12.01-2007 and CSA approvals for Class I, Division 2, Groups A, B, C, D, T6; Class II, Division 2 Groups F, G; Class III.