Propper Blue Autoclavable Tape
Blue Autoclavable Tape provides a means of identifying steam-sterilized packs while ensuring their security during sterilization.
Supplier:  Propper 26810900
 View more versions of this product
Catalog No. 11-889-5
Explore available promotions
Alert:
Cannot be used for ethylene oxide or dry heat sterilization. Do not use on linen or muslin wraps.
Description
Description
  • Specially designed with an aggressive adhesive backing to stick securely to all disposable wraps
  • High-contrast, white, steam-sensitive stripes turn dark when subjected to steam autoclaving temperatures
Order Info
In 60 yd. (54.5m) lengths
Specifications
Specifications
Autoclavable
Synthetic Resin
Tape
Steam Autoclave Tape
Different types of sterilization packs, to distinguish processed and unprocessed items
60 yd. x 1 in.(54.9 m x 25.4 mm)
54.9 m
25.4 mm
7.62 cm
Rubber
White, Blue
Crepe Paper
Non-sterile
1 Ea.
180 ft.
1 in.
3 in.
Product Suggestions
Product Suggestions