missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BOC-L-Alanine, 99+%, Thermo Scientific Chemicals

$233.90 - $233.90

Chemical Identifiers

CAS 15761-38-3
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037225
InChI Key QVHJQCGUWFKTSE-YFKPBYRVSA-N
Synonym boc-ala-oh, n-tert-butoxycarbonyl-l-alanine, boc-l-alanine, n-boc-l-alanine, boc-l-ala-oh, n-t-boc-l-alanine, boc-ala, tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, n-alpha-t-boc-l-alanine
PubChem CID 85082
SMILES C[C@H](NC(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC275490500
SDS View Documents
Thermo Scientific Chemicals
275490500
50 g Glass Bottle
Each for $233.90
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15761-38-3
189.21
QVHJQCGUWFKTSE-YFKPBYRVSA-N
85082
C8H15NO4
MFCD00037225
boc-ala-oh, n-tert-butoxycarbonyl-l-alanine, boc-l-alanine, n-boc-l-alanine, boc-l-ala-oh, n-t-boc-l-alanine, boc-ala, tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, n-alpha-t-boc-l-alanine
C[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

81.0°C to 84.0°C
White
99+%
Glass Bottle
MFCD00037225
04, 2507
QVHJQCGUWFKTSE-YFKPBYRVSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
50 g
85082
≥99%
BOC-L-Alanine
15761-38-3
Authentic
C8H15NO4
(CH3)3COC(O)NHC(CH3)COOH
− 26.00
boc-ala-oh, n-tert-butoxycarbonyl-l-alanine, boc-l-alanine, n-boc-l-alanine, boc-l-ala-oh, n-t-boc-l-alanine, boc-ala, tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, n-alpha-t-boc-l-alanine
C[C@H](NC(=O)OC(C)(C)C)C(O)=O
−26.00 (20.00°C c=2,CH3COOH)
189.21
189.21
Granular Powder
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only