Boiling Chips
Supplier:  Customer Channels Group (Ccg) 501AE
Catalog No. S50414
Explore available promotions
Description
Description
Helps prevent bumping
These inert non-volatile granules resist most chemical action except those of concentrated sulfuric acids and strong alkalies. And, they won't contaminate distillate.
Specifications
Specifications
Porous Carbon
8 oz.