missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mopec Bone Dust Collector for Stryker Saw and Mopec Autopsy Saw

Mopec Bone Dust Collector for Stryker Saw and Mopec Autopsy Saw features Hepa filtration to minimize exposure to particulate matter. Attaches directly to the autopsy saw for safety and efficiency.

$10770.68 - $10840.89

Specifications

Frequency 60 Hz
Voltage 115 V
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
22-444-343
View Documents
Mopec
BD075
Mopec autopsy saw (excluding model 5000) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BD080
View Documents
Mopec
BD080
Stryker saw N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Hepa Vacuum includes Hepa Filter, Vacuum Hose (102in.) and Y Bracket
  • One Year Warranty
  • 115V, 60Hz with US plug

Warranty

1 year

Specifications

Specifications

60 Hz
115 V
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'