missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Borane-2-methylpyridine complex, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6664206

 View more versions of this product

Catalog No. AAH6664206


Explore available promotions

Description

Description

Used as reactant for N-Benzyl-protection of amino acid derivatives by reductive alkylation, reductive alkoxyamination, as reducing agent for the labeling of oligosaccharides by reductive amination, synthesis of alkoxyamine derivatives, via reduction of oxime ethers, reductive amination reactions of C1-C10 aldehyde 2,4-dinitrophenylhydrazones, synthesis of various trifluoromethylated amino compounds and an alternative reagent for reductive aminations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as reactant for N-Benzyl-protection of amino acid derivatives by reductive alkylation, reductive alkoxyamination, as reducing agent for the labeling of oligosaccharides by reductive amination, synthesis of alkoxyamine derivatives, via reduction of oxime ethers, reductive amination reactions of C1-C10 aldehyde 2,4-dinitrophenylhydrazones, synthesis of various trifluoromethylated amino compounds and an alternative reagent for reductive aminations.

Solubility
Slightly sol. in water, cyclohexane, very sol. in methanol, THF, diglyme, and benzene

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Borane-2-methylpyridine complex
44°C to 50°C
100°C (212°F)
MFCD07784361
UN2813
2-Picoline borane complex
ZMASBEFBIXMNCP-UHFFFAOYSA-N
2-methylpyridine borane
106.96
Solid
3999-38-0
White
C6H10BN
5 g
Moisture sensitive
Slightly sol; in water,cyclohexane,very sol; in methanol,THF,diglyme,and benzene
B.CC1=CC=CC=N1
106.96
95%

Chemical Identifiers

3999-38-0
106.96
ZMASBEFBIXMNCP-UHFFFAOYSA-N
2-methylpyridine borane
C6H10BN
MFCD07784361
2-Picoline borane complex
B.CC1=CC=CC=N1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P223-P231+P232-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P335+P334-P362-P370+P378q-P501c

H261-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, N.O.S.

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only