Learn More
Borane-methyl sulfide complex, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 185611000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Borane-methyl sulfide complex | |
1M Solutionin Methylene Chloride | |
C2H9BS | |
MFCD00013189 | |
1.2800g/mL | |
dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms | |
RMHDLBZYPISZOI-UHFFFAOYSA-N | |
boron;methylsulfanylmethane | |
9833925 | |
AcroSeal™ Glass bottle | |
100 mL | |
Liquid |
Colorless | |
13292-87-0,75-09-2 | |
(CH3)2S·BH3 | |
0.95 to 1.05M | |
15, 1336 | |
Solubility in water: reacts. | |
B.CSC | |
75.96 | |
75.95 | |
1.28 | |
18°C |
Chemical Identifiers
13292-87-0 | |
75.96 | |
RMHDLBZYPISZOI-UHFFFAOYSA-N | |
9833925 |
C2H9BS | |
MFCD00013189 | |
dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms | |
B.CSC |
Safety and Handling
GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously. Harmful if swallowed. Causes skin irritation. Causes serious eye damage. May cause drowsiness or dizziness. Suspected of causing cancer if inhaled. Reacts violently with water.
GHS P Statement
Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Danger
EINECSNumber : 236-313-6
RTECSNumber : PV5080000
TSCA : TSCA
RUO – Research Use Only