Learn More
Borane-N,N-diethylaniline complex, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 176771000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Borane-N, N-diethylaniline complex | |
-30°C to 27°C | |
0.9200g/mL | |
Authentic | |
AcroSeal™ Glass bottle | |
1.5245 to 1.5265 | |
MFCD00013187 | |
0.92 | |
Solubility in water: may decompose | |
[B].CCN(CC)C1=CC=CC=C1 | |
160.05 | |
163.07 | |
Liquid |
13289-97-9,91-66-7 | |
Colorless to Amber | |
63°C | |
97% | |
C10H15BN | |
C6H5N(C2H5)2·BH3 | |
100 g | |
n,n-diethylaniline borane, borane-n,n-diethylaniline complex, borane-n,n-diethylaniline, n,n-diethylanilineborane, diethylaniline borane, n,n-diethylaniline trihydroboron, n.n diethylanilineborane, boron; n,n-diethylaniline, boron, n,n-diethylbenzenamine trihydro-, t-4, borane n,n-diethylaniline complex | |
KHYAFFAGZNCWPT-UHFFFAOYSA-N | |
boron;N,N-diethylaniline | |
6335292 | |
97% |
Chemical Identifiers
13289-97-9 | |
160.05 | |
KHYAFFAGZNCWPT-UHFFFAOYSA-N | |
6335292 | |
[B].CCN(CC)C1=CC=CC=C1 |
C10H15BN | |
MFCD00013187 | |
n,n-diethylaniline borane, borane-n,n-diethylaniline complex, borane-n,n-diethylaniline, n,n-diethylanilineborane, diethylaniline borane, n,n-diethylaniline trihydroboron, n.n diethylanilineborane, boron; n,n-diethylaniline, boron, n,n-diethylbenzenamine trihydro-, t-4, borane n,n-diethylaniline complex | |
boron;N,N-diethylaniline |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
In contact with wat
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
EINECSNumber : 236-305-2
RUO – Research Use Only