Learn More
Borane-tert-butylamine complex, 95%, powder, Thermo Scientific Chemicals
$110.47 - $336.69
Chemical Identifiers
CAS | 7337-45-3 |
---|---|
Molecular Formula | C4H14BN |
Molecular Weight (g/mol) | 86.97 |
MDL Number | MFCD00075635 |
InChI Key | GKFJEDWZQZKYHV-UHFFFAOYSA-N |
Synonym | tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane |
PubChem CID | 6364547 |
IUPAC Name | boron;2-methylpropan-2-amine |
SMILES | B.CC(C)(C)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC177300250
|
Thermo Scientific Chemicals
177300250 |
25 g | Glass Bottle |
Each for $110.47
|
|
||||
AC177301000
|
Thermo Scientific Chemicals
177301000 |
100 g | Glass bottle |
Each for $336.69
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
7337-45-3 | |
86.97 | |
GKFJEDWZQZKYHV-UHFFFAOYSA-N | |
6364547 | |
B.CC(C)(C)N |
C4H14BN | |
MFCD00075635 | |
tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane | |
boron;2-methylpropan-2-amine |
Specifications
7337-45-3,75-64-9 | |
White | |
95% | |
C4H14BN | |
MFCD00075635 | |
11,87 | |
Solubility in water: 2.7g/100mL (20°C) | |
B.CC(C)(C)N | |
86.97 | |
86.97 | |
Fine Crystalline Powder |
96°C to 101°C | |
Authentic | |
Glass Bottle | |
(CH3)3CNH2·BH3 | |
25 g | |
tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane | |
GKFJEDWZQZKYHV-UHFFFAOYSA-N | |
boron;2-methylpropan-2-amine | |
6364547 | |
95% | |
Borane-tert-butylamine complex, 95% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Toxic in contact with skin.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Flammable solid.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume
GHS Signal Word: Danger
EINECSNumber : 230-851-5
RTECSNumber : EO3900000
TSCA : TSCA
RUO – Research Use Only