missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, 99.5%, for analysis

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol

Supplier:  Thermo Scientific Chemicals 217085000

 View more versions of this product

Catalog No. AC217085000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer, Lewis acid
Specifications

Specifications

3.6 to 4.0 (4% at 20°C)
White
169.0°C
5ppm max.
Plastic bottle
500 g
Fine Crystals
MFCD00011337
15, 1339
Solubility in water: 49.5g/L (20°C). Other solubilities: 1g/18mL cold alcohol - 1g/6mL boiling alcohol, 1g/4mL glycerol, nacl, licl, mineral acids decrease the solubility, kno3,k2so4,kcl,nano3,na2so4 increase the sol.
OB(O)O
61.83
CHEBI:33118
99.5%
Boric acid
p.a.
10043-35-3
0.05% max.
99.5%
BH3O3
H3BO3
01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
7628
61.83
Analytical

Chemical Identifiers

10043-35-3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
7628
boric acid
BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
CHEBI:33118
OB(O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement May damage fertility. May damage the unborn child.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 233-139-2

RTECSNumber : ED4550000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only