missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals

$110.46 - $1369.12

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC315181000
SDS View Documents
Thermo Scientific Chemicals
315181000
100 g Plastic bottle
Each for $110.46
 
AC315185000
SDS View Documents
Thermo Scientific Chemicals
315185000
500 g Plastic bottle
Each for $335.27
 
AC315180025
SDS View Documents
Thermo Scientific Chemicals
315180025
2.5 kg Plastic bottle
Each for $1,369.12
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis, Lewis acid
Specifications

Chemical Identifiers

10043-35-3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
7628
boric acid
BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
CHEBI:33118
OB(O)O

Specifications

White
10043-35-3
99.99%
BH3O3
H3BO3
01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
7628
61.83
Trace Metal Basis
169.0°C
Plastic bottle
100 g
Crystals or Crystalline Powder
MFCD00011337
15, 1339
Solubility in water: 49.5g/L (20°C). Other solubilities: 1g/18mL cold alcohol - 1g/6mL boiling alcohol, 1g/4mL glycerol, nacl, licl, mineral acids decrease the solubility, kno3,k2so4,kcl,nano3,na2so4 increase the sol.
OB(O)O
61.83
CHEBI:33118
99.99%
Boric acid, Extra Pure
Videos
Safety and Handling

Safety and Handling

GHS P Statement May damage fertility. May damage the unborn child.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only