missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boric Acid, Crystal, ACS, 99.5%, Spectrum™ Chemical
$246.97 - $1811.47
Chemical Identifiers
CAS | 10043-35-3 |
---|---|
Molecular Formula | BH3O3 |
Molecular Weight (g/mol) | 61.83 |
MDL Number | MFCD00011337 |
InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
IUPAC Name | boric acid |
SMILES | OB(O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18609162
|
Spectrum Chemical Mfg Cor
B1130125GM |
125 g | Poly Bottle |
Each for $246.97
|
|
||||
18609164
|
Spectrum Chemical Mfg Cor
B1130500GM |
500 g | Poly Bottle |
Each for $397.97
|
|
||||
18609166
|
Spectrum Chemical Mfg Cor
B113025KG |
2.5 kg | Poly Bottle |
Each for $1,048.97
|
|
||||
18609163
|
Spectrum Chemical Mfg Cor
B113012KG |
12 kg | Poly Pail |
Each for $1,811.47
|
|
||||
Chemical Identifiers
10043-35-3 | |
61.83 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
OB(O)O |
BH3O3 | |
MFCD00011337 | |
boric acid |
Specifications
10043-35-3 | |
1e-05 | |
125 g | |
MFCD00011337 | |
OB(O)O | |
61.83 | |
ACS |
1 | |
Poly Bottle | |
BH3O3 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
boric acid | |
99.5% |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.