missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boric Acid (Crystalline/NF/EP/BP/JP), Fisher Chemical™
Supplier: Fisher Chemical A7912
Specifications
3.8 to 4.8 (EP) | |
White | |
169°C | |
≤15ppm (BP, EP), ≤10ppm (JP), 10ppm max. (JP), 15% max. (EP), 15ppm max. (BP) | |
Pass Test (NF, BP, EP, JP), Pass Test (EP), Pass Test (JP), Pass Test (NF), Pass Test (NF, BP, EP | |
2.7mbar at 20°C | |
BH3O3 | |
MFCD00011337 | |
Solubility in water: partially soluble | |
OB(O)O | |
61.83 | |
CHEBI:33118 | |
≥99.5% (JP) |
Boric Acid | |
Clear and Colorless (BP), Clear and colorless (BP, EP), Clear and Colorless (EP) | |
10043-35-3 | |
0.002% max. (NF), 0.002% max. (NF), 15ppm max. (BP), 15% max. (EP) 10ppm max. (JP) | |
Poly Pail | |
12 kg | |
Powder/Solid | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
boric acid | |
7628 | |
61.83 | |
BP/EP/JP/NF |
Chemical Identifiers
10043-35-3 | |
61.83 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
7628 | |
boric acid |
BH3O3 | |
MFCD00011337 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
CHEBI:33118 | |
OB(O)O |
Safety and Handling
WARNING!
Emergency Overview
May impair fertility. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Ingestion may cause gastrointestinal irritation, nausea, vomiting and diarrhea. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.
NFPA
Health:2
Flammability:0
Instability:1