missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boric acid, Electrophoresis Grade, 99.5% min, Thermo Scientific Chemicals
$103.00 - $509.51
Chemical Identifiers
CAS | 10043-35-3 |
---|---|
Molecular Formula | BH3O3 |
Molecular Weight (g/mol) | 61.83 |
MDL Number | MFCD00011337 |
InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
Synonym | orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe |
PubChem CID | 7628 |
ChEBI | CHEBI:33118 |
IUPAC Name | boric acid |
SMILES | OB(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6720236
|
Thermo Scientific Chemicals
J6720236 |
500 g |
Each for $103.00
|
|
|||||
AAJ67202A1
|
Thermo Scientific Chemicals
J67202A1 |
1 kg |
Each for $152.48
|
|
|||||
AAJ67202A7
|
Thermo Scientific Chemicals
J67202A7 |
5 kg |
Each for $509.51
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10043-35-3 | |
61.83 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
7628 | |
boric acid |
BH3O3 | |
MFCD00011337 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
CHEBI:33118 | |
OB(O)O |
Specifications
3.8 to 4.8 | |
∼185°C (decomposition) | |
1.435 g/mL | |
500 g | |
Powder | |
1697939 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
OB(O)O | |
61.83 | |
CHEBI:33118 | |
≥99.5% | |
Boric acid |
White | |
10043-35-3 | |
Odorless | |
BH3O3 | |
MFCD00011337 | |
14,1336 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
boric acid | |
7628 | |
61.83 | |
Electrophoresis |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P260-P281-P308+P313-P501c
H360FD
EINECSNumber : 233-139-2
RTECSNumber : ED4550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only