missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boric Acid, Granular, N.F., J.T. Baker™

Supplier:  Avantor J.T.Baker 009105

 View more versions of this product

Catalog No. 02002448


Explore available promotions

Description

Description

Specifications

Specifications

Boric Acid
Poly Bottle
2.5 kg
Granular
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
OB(O)O
61.83
CHEBI:33118
NF
10043-35-3
1L = 1.43kg
BH3O3
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
7628
61.83

Chemical Identifiers

10043-35-3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
7628
boric acid
BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
CHEBI:33118
OB(O)O
Videos
SDS
Documents

Documents

Promotions

Promotions