missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boric Acid, Granular, Ultrapure Bioreagent, J.T. Baker™
High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography
Supplier: Avantor 403505
Description
- For Molecular Biology Buffers
Specifications
Boric Acid | |
Poly Bottle | |
2.5 kg | |
Granular | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
OB(O)O | |
61.83 | |
CHEBI:33118 | |
Ultrapure Bioreagent |
10043-35-3 | |
1L = 1.43kg | |
BH3O3 | |
MFCD00011337 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
boric acid | |
7628 | |
61.83 |
Chemical Identifiers
10043-35-3 | |
61.83 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
7628 | |
boric acid |
BH3O3 | |
MFCD00011337 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
CHEBI:33118 | |
OB(O)O |