missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boric Acid (Powder/Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical FLA7410

 View more versions of this product

Catalog No. A74-10


Explore available promotions

Description

Description

Specifications

Specifications

3.8 to 4.8
White
10043-35-3
Pass Test
Poly Pail
≥99.5 %
BH3O3
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
7628
61.83
Certified ACS
Boric Acid
169°C
0.001% max.
0.05% max.
2.7mbar at 20°C
10 kg
Powder/Solid
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
OB(O)O
61.83
CHEBI:33118
≥99.5%

Chemical Identifiers

10043-35-3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
7628
boric acid
BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
CHEBI:33118
OB(O)O
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
May impair fertility. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Ingestion may cause gastrointestinal irritation, nausea, vomiting and diarrhea. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:2
Flammability:0
Instability:1

SDS Documents

Documents

Promotions

Promotions