missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., buffer substance, ≥99.8%, Honeywell Fluka™

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., buffer substance,≥99.8%

$206.15 - $2575.75

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002311
View Documents
Honeywell Chemicals
31146500G
500 g
Each for $206.15
 
6002309
View Documents
Honeywell Chemicals
311461KG
1 kg
Each for $401.78
 
6002310
View Documents
Honeywell Chemicals
3114625KG
2.5 kg
Each for $547.23
 
60045920
View Documents
Honeywell Chemicals
311466X1KG
6 x 1 kg
Case of 6 Each for $2,121.05
 
60045921
View Documents
Honeywell Chemicals
311466X25KG
6 x 2.5 kg
Case of 6 Each for $2,575.75
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

10043-35-3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
7628
boric acid
BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
CHEBI:33118
OB(O)O

Specifications

10043-35-3
500 g
H3BO3
NONH for all modes of transport
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
7628
61.83g/mol
ACS/puriss. p.a.
2.6mmHg (20°C)
BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
OB(O)O
61.83
CHEBI:33118
≥99.8%
Boric Acid, Reag. Ph. Eur. reag. ISO
Videos
Safety and Handling

Safety and Handling

P201-P280-P308 + P313

H360FD

EINECSNumber : 233-139-2

RTECSNumber : ED4550000

SDS
missing translation for 'documents'

missing translation for 'documents'