missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boron nitride paste, Thermo Scientific Chemicals
Has Resistance Against Molten Metals and Salts, High thermal Conductivity and Dielectric Strength
Supplier: Thermo Scientific Chemicals 04377336
Description
Resistance against molten metals and salts, high thermal conductivity and dielectric strength
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boron nitride | |
10043-11-5 | |
500 g | |
MFCD00011317 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
boranimine | |
66227 | |
Odorless | |
1.4 |
White | |
Paste | |
BN | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
B=N | |
24.82 | |
CHEBI:50883 | |
White paste |
Chemical Identifiers
10043-11-5 | |
24.82 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
66227 | |
B=N |
BN | |
MFCD00011317 | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
CHEBI:50883 |
Safety and Handling
H319-H335: Causes serious eye irritation. May cause respiratory irritation.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only