missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron oxide, 97.5% min, typically 98.5%, Thermo Scientific Chemicals

$93.30 - $710.38

Chemical Identifiers

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
PubChem CID 518682
ChEBI CHEBI:30163
SMILES O=BOB=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1229036
View Documents
Thermo Scientific Chemicals
01229036
500 g
Each for $93.30
 
AA12290A3
View Documents
Thermo Scientific Chemicals
012290A3
2 kg
Each for $206.36
 
AA12290A9
View Documents
Thermo Scientific Chemicals
012290A9
10 kg
Each for $710.38
 
Description

Description

It is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride, and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from Talc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1303-86-2
69.62
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
518682
O=BOB=O
B2O3
MFCD00011315
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
CHEBI:30163

Specifications

1860°C
1303-86-2
97.5% min., typically 98.5%
MFCD00011315
Slightly soluble in water and methanol.
O=BOB=O
69.62
CHEBI:30163
Hygroscopic
Boron oxide
∽450°C
500 g
B2O3
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
oxo[(oxoboranyl)oxy]borane
518682
69.62
2.46 ^+ 0.01 g/mL
Videos
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H360FD

EINECSNumber : 215-125-8

RTECSNumber : ED7900000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only