Learn More
Boron powder, crystalline, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
$648.78 - $2733.71
Chemical Identifiers
CAS | 7440-42-8 |
---|---|
Molecular Formula | B |
Molecular Weight (g/mol) | 10.81 |
MDL Number | MFCD00134034 MFCD00151272 |
InChI Key | ZOXJGFHDIHLPTG-UHFFFAOYSA-N |
Synonym | boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne |
PubChem CID | 5462311 |
ChEBI | CHEBI:27560 |
IUPAC Name | boron |
SMILES | [B] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-42-8 | |
10.81 | |
ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
5462311 | |
boron |
B | |
MFCD00134034 MFCD00151272 | |
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne | |
CHEBI:27560 | |
[B] |
Specifications
7440-42-8 | |
MFCD00134034 MFCD00151272 | |
1 g | |
ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
boron | |
5462311 | |
99.9999% | |
−4+40 Mesh | |
Crystalline Powder | |
(metals basis) |
B | |
UN3178 | |
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne | |
[B] | |
10.81 | |
CHEBI:27560 | |
Puratronic™ | |
Odorless | |
Boron powder |
Safety and Handling
GHS H Statement
H228-H302-H332-H335
Flammable solid.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber : 231-151-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only