missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron tribromide dimethyl sulfide, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

$226.21 - $226.21

Specifications

Chemical Name or Material Boron tribromide dimethyl sulfide
Name Note 1M solution in methylene chloride
CAS 29957-59-3,75-09-2
Molecular Formula C2H6BBr3S
MDL Number MFCD00043296
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC428321000
View Documents
Thermo Scientific Chemicals
428321000
100 mL AcroSeal™ Glass bottle
Each for $226.21
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Boron tribromide dimethyl sulfide
29957-59-3,75-09-2
MFCD00043296
Solubility in water: reacts
CSC.BrB(Br)Br
312.65
312.66
1.4560g/mL
1M solution in methylene chloride
C2H6BBr3S
tribromoborane-methyl sulfide, boron tribromide-methyl sulfide complex, boron tribromide dimethyl sulfide complex, tribromo dimethyl-??-sulfanylidene-??-borane, dimethyl sulfide-tribromoborane, tribromo dimethylsulfonio boranuide, boron tribromide-methylsulfide complex, borontribromide-methyl sulfide complex, boron tribromide dimethyl sulfide complex solution
YNLFEVAOQLXINF-UHFFFAOYSA-N
(methylsulfanyl)methane; tribromoborane
4181510
1.456
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only