missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
$108.33 - $602.55
Specifications
Chemical Name or Material | Boron trichloride |
---|---|
Color | Colorless to Yellow |
Name Note | 1M Solution in Hexane |
CAS | 10294-34-5,110-54-3 |
Molecular Formula | BCl3 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC198921000
|
Thermo Scientific Chemicals
198921000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $108.33
|
|
||||
AC198928000
|
Thermo Scientific Chemicals
198928000 |
800 mL | AcroSeal™ Glass bottle |
Each for $602.55
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Boron trichloride | |
1M Solution in Hexane | |
BCl3 | |
MFCD00011313 | |
01,67; 02,34; 03,31; 04,42; 05,50; 06,65; 09,62; 13,43; 15,44 | |
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas | |
FAQYAMRNWDIXMY-UHFFFAOYSA-N | |
trichloroborane | |
25135 | |
0.738 | |
Solution |
Colorless to Yellow | |
10294-34-5,110-54-3 | |
BCl3 | |
0.9 to 1.1M | |
0.7380g/mL | |
Solubility in water: may decompose. | |
ClB(Cl)Cl | |
117.16 | |
117.17 | |
−17°C |
RUO – Research Use Only